Thermocouple Adapter, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This thermocouple adapter has a lower extension to better help support the thermocouple.
- With lower extension
- Compression-style Ace-Thred nylon bushing
- PTFE ferrule
Designed for use when thermocouples are bending or appearing to move excesivelly within a joint. The thermocouple is secured with a compression style Ace-Thred nylon bushing and PTFE ferrule.
Customers Who Bought This Also Bought