KIMBLE® Hose Inlet Adapter with Fritted Disc, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Adapter has a 170-220 micron porosity disc between the hose connection and the inner joint.
- Fritted disc between joint and hose connection
- Inner [ST] joint
- Manufactured from borosilicate glass
Fritted disc prevents solids from being introduced into the system.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought