BRAND PCR Plates for Roche® LightCycler® 480, BrandTech®
Fournisseur: Brandtech
Certificats
A propos de cet article
These white PCR plates provide high signal to noise for real-time/qPCR.
- Uniformally coloured with titanium dioxide (TiO₂)
- Smooth surfaces to optimise reflection of the fluorescence signal
- Extra-thin wall thickness to provide optimal thermal transfer and short cycle times
- 96-well plates have black alphanumeric code, no additional markings on the cut corner
- Manufactured under modern cleanroom conditions according to ISO 14 644-1 class 5 to 8
- DNase-, DNA- and RNase-free
Available with 96-well low profile and 384-well formats. Both plates are compatible with many thermal cyclers, but are perfectly suited for use in the Roche® LightCycler® 480.
These BRAND PCR Plates are made of PP.
Customers Who Bought This Also Bought