KIMBLE® Straight Connecting Adapters, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Straight Connecting Adapters expand the length of an apparatus, and are also useful as an air condenser.
- 155mm and 160mm sizes have an outer [ST] joint at one end and inner [ST] joint on the other end
- 110mm size has inner [ST] joints at both ends
- Manufactured from borosilicate glass
The 155mm and 160mm adapters feature outer Standard Taper joints at one end and inner Standard Taper joints on the other end
Adapters are manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
The 110mm adapter has inner Standard Taper joints at both ends.
Customers Who Bought This Also Bought