KIMBLE® PTFE Threaded Microscale Connecting Adapters with [ST] Inner Joint, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Adapters allow interfacing between outer Standard Taper jointed glassware with threaded microscale components.
- Threaded on one end
- With [ST] inner joint
- Manufactured from PTFE
Provides a shorter travel path than the equivalent glass connector.
Customers Who Bought This Also Bought