PTFE Plug-Style Stopcocks, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
Certificats
A propos de cet article
PTFE stopcock plug has a 1:5 taper and is chemically-resistant and self-lubricating.
Customers Who Bought This Also Bought
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Fournisseur: VWR International
541,80 $
Nalgene® Laboratory Notebooks, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
172,57 $ to 531,06 $
SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
362,33 $
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
570,32 $
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
464,49 $
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Fournisseur: Saint Gobain Performance Plastics
1 556,03 $
Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
534,61 $ to 1 523,31 $
Nalgene® PolyPaper® Laboratory Notebooks, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
397,26 $ to 1 244,71 $