Science Take-Out® Genetic Engineering: Way To Grow
Fournisseur: SCIENCE TAKE-OUT, LLC
Certificats
A propos de cet article
How Does Genetic Engineering Produce Human Growth Hormone?
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
How can genetic engineering be used to help a child who is not growing normally? Conduct simulated tests to determine what is causing the child's slow growth. Learn how hormone replacement therapy is used to treat human growth hormone deficiency. Model how genetic engineering can be used to create bacteria that produce human growth hormone. This complete experiment setup contains all materials and does not require lab equipment.
Customers Who Bought This Also Bought