Adsorbent Paper, Apacor Limited
# de catalogue 10144-692
Fournisseur: Apacor
Spécifications
- Dimensions:7.6 x 15.2 cm (3 x 6")
- Description:Adsorbent Paper
- Cat. no.:10144-692
- Supplier No.:82600
Spécifications
A propos de cet article
Adsorbent paper is designed for use in blotting a sample before media application.
The papers are made from a special non-leaching high-adsorbency material, which allows fast preparation of the electrophoresis media.
Certifications: CE certified.
Customers Who Bought This Also Bought