Cellulose Acetate Membrane, Apacor
Fournisseur: Apacor
Certificats
A propos de cet article
Membranes are designed for use in the electrophoretic separations of serum proteins.
Easy to handle membranes offer high sensitivity, clear resolution, and reproducible results.
Certifications: CE certified.
Ordering information: Available in a range of membrane sizes.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.