Display Bottle, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Used to display materials, particularly at exhibits, museums and medical schools
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Finished with uniform walls and a heavy flared base which does not exceed the jar diameter. Specially fitted corks are included and packed separately.
Customers Who Bought This Also Bought