Gas Washing Bottle, Fritted, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Large capacity, low-form gas washing bottle has a 34/28 standard taper top outer joint.
Filter candle is 25 Ext.Ø×70 mm long, allowing a large gas flow to be dispersed throughout the absorbing material.
Ordering information: Complete bottle is supplied with one each size 34 Keck clamp. Available with a coarse or extra coarse porosity filter candle.
Customers Who Bought This Also Bought