Nalgene® Vacuum Gasket for Filter Funnels, Thermo Scientific
Catalog # 28198-898
Supplier: Thermo Fisher Scientific
Specifications
- Description:Nalgene® vacuum gasket, TPE, 2.76 cm (1.09") height, 3.96 cm (1.56") OD
- Cat. no.:28198-898
- Supplier No.:DS0395-0708
Specifications
About this item
Replace No.7 or No. 8 stoppers in any manifold or filtering flask with the Nalgene™ Vacuum Gasket for Filter Funnels.
Thermoplastic elastomer forms a reliable seal, and permits filter funnel to slide quickly and easily in and out of the gasket. These filtration products offer reliability, accuracy, and safety for many research applications.
Frequently Bought Together