A propos de cet article
The DATK32 antibody is specific for a combinatorial epitope of the mouse integrin alpha 4/beta 7 (LPAM-1) heterodimer. LPAM-1 is expressed on most mature lymphocytes and sub-populations of thymocytes and bone marrow cells. It is involved in lymphocyte adhesion and migration and is a receptor for several extracellular matrix proteins including VCAM-1, fibronectin, and MAdCAM-1.
DATK32 is reported to induce LPAM-1 dependent lymphocyte aggregation and block adhesion to VCAM-1, fibronectin, and MAdCAM-1.
Type: Primary
Antigen: LPAM-1
Clonality: Monoclonal
Clone: DATK32
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 10050-778
- 10050-776
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- DATK32
- DATK32
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Isotype:
- IgG2a
- IgG2a
- Neutralisation:
- Yes
- Yes