20-Place Cardboard Slide Trays
Supplier: Heathrow Scientific
Certificates
About this item
Cardboard slide folders offer a convenient solution for safely working, transporting and short-term storing of slides.
- Compartment holder is made from a double layer of densely pressed cardboard
- Cover is single layer cardboard
- Front protection flaps are attached with durable tape
- Recessed slide compartments and inside covers for specimen protection
- Slides cannot be accessed when cover is closed reducing contamination
Index on the front cover corresponds with the number grid inside for identification. Cardboard slide trays hold standard slides 76×26 mm (3×1").
Frequently Bought Together