Mega Tube Racks for 13 - 16 mm Tubes
Supplier: Heathrow Scientific
Certificates
About this item
Fully-assembled, Mega tube racks are for high throughput labs requiring large sample storage.
- Sturdy and light weight material for easy and secure transport of blood collection tubes
- Alphanumeric identification for quick location and retrieval of tubes
- Place different test groupings into one rack and eliminate need for multiple tube racks
Racks are manufactured from polypropylene and are autoclavable.
High-capacity storage for 10 ml tubes.
Frequently Bought Together