Brooms, Soft, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
This soft/split bristle floor broom is suitable for dry environments, and is ideal for sweeping flour and starch as well as other dry powders
Block material is polypropylene; bristle material is polyester.
Certifications: FDA Compliant Raw Material.
Customers Who Bought This Also Bought