ContactUsLinkComponent
Nunc® Conical Centrifuge Tube, 250 ml, Thermo Scientific
Nunc® Conical Centrifuge Tube, 250 ml, Thermo Scientific
# de catalogue 10147-000
Conical centriguge tube, polypropylene, sterile, 250mL, 4 per pack, 40 per case
Nunc® Conical Centrifuge Tube, 250 ml, Thermo Scientific
# de catalogue 10147-000
Spécifications
  • Sterility:
    Sterile
  • Volume:
    250 ml
  • Material:
    PP
  • Packaging:
    40 Tubes/Case
  • Supplier No.:
    376814
Spécifications
A propos de cet article
The Nunc® 250 ml conical centrifuge Tube offers added capacity for larger samples with a one-of-a-kind wide mouth design.
  • RNase- and DNase-free
It provides open, easy access for pipetting, decanting, and removal of the pellet following centrifugation.
Certifications: USP Class VI.
Packaging: Tubes are packaged 40 per case.
Customers Who Bought This Also Bought