ContactUsLinkComponent
Tissue Cassette Storage Cabinet, Boekel
Tissue Cassette Storage Cabinet, Boekel
# de catalogue 10144-720
Fournisseur:  Boekel
CABINET TISSUE CASSETTE STORAGE
Tissue Cassette Storage Cabinet, Boekel
# de catalogue 10144-720
Fournisseur:  Boekel
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Tissue cassette storage cabinet
  • Material:
    Epoxy powder-coated steel
  • Cat. no.:
    10144-720
  • Supplier No.:
    143000
Spécifications
A propos de cet article
A robust, modular storage solution for formalin fixed paraffin embedded tissue samples.
  • Economical option for storing cassettes
  • Foam organizers included
  • Easy slide drawers with customizable labels
Each drawer includes easily removable crystal clear trays along with foam organizers for keeping cassettes upright.
Store up to 1200 cassettes in the easy slide drawers, each with customizable labels. The non-tip design allows for stacking of up to 3 cabinets high.
Customers Who Bought This Also Bought