Women's Button Lab Coats, 80/20 Poly-Cotton
Fournisseur: VWR International
Certificats
A propos de cet article
These fashionable lab coats for women feature a five-button front closure, tapered back panel with a half belt, and are made with a soft but very durable 80/20 poly-cotton fabric material blend for long-term wear that can stand up to industrial laundering.
- Industrial laundry friendly
- Durable and comfortable
- Hemmed cuffs
- Side vents for easy access to pant pockets
- Designed specifically for women, this coat is tapered and has a princess-belt back
Overall length is 91.4 cm (36"). Lab coats feature a plain front, with no embroidery. Available in white only.
Designed to provide a clean professional appearance, they have 3 pockets; one breast pocket with a pencil slot, and two lower hip pockets large enough for a mini-tablet. For added convenience, there are side vents to access pants pockets.
Ordering information: Sizes 3X-Large through 5X-Large require extended lead time. Contact your Avantor sales representative for more information.
Spécifications
- Catalog No:
- 10120-354
- 10120-356
- 10120-358
- 10120-360
- 10120-362
- 10120-344
- 10120-346
- 10120-348
- 10120-350
- 10120-352
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Gender:
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 80/20 Poly-cotton
- 80/20 Poly-cotton
- 80/20 Poly-cotton
- 80/20 Poly-cotton
- 80/20 Poly-cotton
- 80/20 Poly-cotton
- 80/20 Poly-cotton
- 80/20 Poly-cotton
- 80/20 Poly-cotton
- 80/20 Poly-cotton