Pierceable Plug Stopper - For 13 mm Tubes, Globe Scientific
These pierceable plug stoppers are intended for recapping vacuum blood tubes and test tubes.
- Thermoplastic elastomer (TPE)
- Assorted colors available
- Fits both glass and plastic tubes
The caps are produced from thermoplastic elastomer (TPE) which can easily be pierced with a syringe.
Ordering information: Stoppers are packaged 1000 per bag.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.