To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Control
- Clone:C1.18.4
- Host:Mouse
- Isotype:IgG2a
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Storage temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested applications:Functional Assay
- Cat. no.:10141-218
- Supplier No.:70-4724-U100
Spécifications
A propos de cet article
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.
Type: Control
Antigen: C1.18.4
Clonality:
Clone: C1.18.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0