A propos de cet article
The 10F.9G2 antibody is specific for mouse CD274, more commonly known as PD-L1 or B7-H1, which acts as a ligand for the T cell co-regulatory receptor PD-1 (CD279). This interaction modulates T cell antigen receptor (TCR) signaling and therefore T cell activation. PD-L1 binding to PD-1 expressed on CD4- CD8- thymocytes participates in the processes of clonal selection, elimination of autoreactive lymphocytes, and development of tolerance. PD-L1 may also bind PD-1 following the receptor’s inducible expression on activated, mature T cells, where it has been proposed to limit T cell activation. PD-L1 is one of a group of “B7” ligands whose interactions with the CD28 receptor family, also including CTLA-4 (CD152), provide a balance of co-stimulatory /co-inhibitory signaling important in T cell activation, tolerance, and autoimmunity.
The 10F.9G2 antibody may be used as a marker for PD-L1 expression on T and B cells, NK cells and on dendritic cells. It is also widely used for analysis of receptor-ligand interaction and function(s) in vitro and in vivo.
Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 10F.9G2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 10051-296
- 10051-294
- Antigen name:
- CD274 molecule
- CD274 molecule
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 10F.9G2
- 10F.9G2
- Host:
- Rat
- Rat
- Gene ID:
- 60533
- 60533
- Isotype:
- IgG2b
- IgG2b