Order Entry
ContactUsLinkComponent
Rat IgG2b Monoclonal Isotype Control [clone: LTF-2] (violetFluor® 450)
Rat IgG2b Monoclonal Isotype Control [clone: LTF-2] (violetFluor® 450)
# de catalogue 10051-848
Fournisseur:  Tonbo Biosciences
Rat IGG2B Isotype control 100UG
Rat IgG2b Monoclonal Isotype Control [clone: LTF-2] (violetFluor® 450)
# de catalogue 10051-848
Fournisseur:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    LTF-2
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Neutralisation:
    Yes
  • Flow cytometry:
    Yes
  • Size:
    100 µg
  • Storage temperature:
    Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested applications:
    Flow Cytometry
  • Cat. no.:
    10051-848
  • Supplier No.:
    75-4031-U100

Spécifications

A propos de cet article

The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.

Type: Control
Antigen: LTF-2
Clonality: Monoclonal
Clone: LTF-2
Conjugation: violetFluor® 450
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:

Customers Who Bought This Also Bought