Industrial Single-Wall Storage Cabinets, Securall
Fournisseur: Securall Cabinets
Certificats
A propos de cet article
Rugged, all welded, 18-gauge steel constructed storage cabinet will provide years of service in both factory and office environments.
- Three-point self-latching doors add even more security
- Shipped fully assembled and ready to use
Double wall cabinet doors feature full length piano hinges and a lockable flush mounted door handle for obstruction free, finger tip operation. Each shelf is easily adjustable and carries a 500 lbs. load capacity.
Spécifications
- Catalog No:
- 14227-664
- 14227-666
- 14227-662
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified