Science Take-Out® Diagnosing Diabetes
Supplier: SCIENCE TAKE-OUT, LLC
Certificates
About this item
Does Your Patient Have Type 1 Or Type 2 Diabetes?
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
Analyze simulated blood plasma samples collected during a glucose tolerance test to determine if your patient has Type 1 or Type 2 diabetes. Create a patient information sheet about diabetes and the glucose tolerance test. Test and graph glucose and insulin levels in simulated blood plasma samples. Analyze the test results to determine if the patient has Type 1 or Type 2 diabetes. This complete experiment setup contains all materials and does not require lab equipment.
Customers Who Bought This Also Bought