Science Take-Out® Flower Forensics
Fournisseur: SCIENCE TAKE-OUT, LLC
Certificats
A propos de cet article
Has A Rare, Black Orchid Been Illegally Cloned?
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
The owner of a local flower store has asked you to investigate a possible crime: the theft and illegal cloning of his prized black orchids. Collect and analyze data to compare a rare black orchid with three suspicious black orchids. Compare orchid structures using diagrams of the four orchid flowers. Perform simulations of: chromatography of plant pigments, enzyme assays, restriction enzyme digestion and gel electrophoresis. This complete experiment setup contains all materials and does not require lab equipment.
Customers Who Bought This Also Bought