Science Take-Out® Yeast Respiration
Fournisseur: SCIENCE TAKE-OUT, LLC
Certificats
A propos de cet article
How Does Changing The Type Of Food Affect Yeast Respiration?
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
Does yeast respiration occur more efficiently at warm or cool temperatures? Which food type (honey or table sugar) is better for yeast respiration? Design and conduct your own experiment to answer this question. This complete experiment setup contains all materials and does not require lab equipment.
Customers Who Bought This Also Bought