ContactUsLinkComponent
Autosampler Syringes for Hitachi/LaChrom HPLC, Hamilton
Autosampler Syringes for Hitachi/LaChrom HPLC, Hamilton
# de catalogue 10118-434
Fournisseur:  Hamilton
UL Listed
Autosampler Syringes for Hitachi/LaChrom HPLC, Hamilton
# de catalogue 10118-434
Fournisseur:  Hamilton
UL Listed
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Volume:
    500 µl
  • Model:
    Gastight 1750 (Needle Not Included)
  • Supplier No.:
    0160310
  • Termination:
    Special
Spécifications
A propos de cet article
The LaChrom Elite Autosampler represents a new standard of performance in HPLC
Whether you need high sample capacity, extreme precision, ultra-low carry over, or rapid cycle times between injections, the LaChrom Elite models excel.
Certifications: UL Listed. CE compliant.
Ordering information: Needle not included. Accessories not included.
Customers Who Bought This Also Bought