HandE-Vac handheld aspirator systems, Argos Technologies
Fournisseur: Argos Technologies
Certificats
A propos de cet article
HandE-Vac Handheld Aspirator Systems are used to rapidly remove liquid from cell culture or multi-well plates and dishes.
Systems can be used with pasteur pipettes, plastic aspiration pipettes, or a variety of adapters. Use with standard in-house vacuum source or with M-Vac or E-Vac Aspiration Systems.
Customers Who Bought This Also Bought