PTFE Tube Compression Elbow to Male NPT, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
IPS PTFE Tube Compression Elbow to Male NPT
Body and ferrules made of virgin PTFE. The nut and ferrule-gripper is made of PVDF. Performance at ambient room conditions: 120 PSI for 1/16" fittings linear decreasing to 80 PSI for 3/4" fittings and 60 PSI for fittings larger than 3/4". Performance at elevated temperatures up to 85C/185F: 90 PSI for 1/16" fittings linear decreasing to 60 PSI for 3/4" fittings and 40 PSI for fittings larger than 3/4".
Customers Who Bought This Also Bought