PTFE Tube Compression Reducing Union, Ace Glass Incorporated
Fournisseur: Avantor
Certificats
A propos de cet article
PTFE Tube Compression Reducing Union
Body and ferrules made of virgin PTFE. The nut and ferrule-gripper is made of PVDF. Performance at ambient room conditions is 120 PSI for 1/16" fittings linear decreasing to 80 PSI for 3/4" fittings and 60 PSI for fittings larger than 3/4". Performance at elevated temperatures up to 85°C/185°F: 90 PSI for 1/16" fittings linear decreasing to 60 PSI for 3/4" fittings and 40 PSI for fittings larger than 3/4".
Spécifications
- Catalog No:
- 10099-328
- 10099-330
- 10099-324
- 10099-320
- 10099-322
- 10099-318
- 10099-334
- 10099-336
- 10099-332
- Max. operating pressure:
- Operating temperature:
Spécifications
Customers Who Bought This Also Bought