PTFE Tube Compression Fitting to Male NPT, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
PTFE tube compression fitting to male NPT
Body and ferrules made of virgin PTFE. The nut and ferrule-gripper is made of PVDF. Performance at ambient room conditions is 120psi for 1/16" fittings linear decreasing to 80psi for 3/4" fittings and 60psi for fittings larger than 3/4". Performance at elevated temperatures up to 85°C/185°F is 90psi for 1/16" fittings linear decreasing to 60psi for 3/4" fittings and 40psi for fittings larger than 3/4".
Customers Who Bought This Also Bought