Gaskets, NW Flange, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
NW (DN) flange glass to glass sealing gasket featuring a PTFE carrier and o-ring available in Viton® (–26 to 240 °C) or FVMQ flurosilicone (–60 to 230 °C).
Excellent for deep vacuum work or a maximum of 2 bar pressure.
Frequently Bought Together