Sterile Pair Packed Class 100 Nitrile Gloves, USP797/800 Compliant , TechNiGlove
Fournisseur: Techniglove
Certificats
A propos de cet article
TechNiGlove pair packed sterile ambidextrous 12" gloves are Technicleaned for Class 100 compatibility and sterilized by gamma irradiation to meet AAMI and ASTM standards.
- 12" ambidextrous, sterile
- L/R wallet pair packed
- TechNiCleaned, USP797/800 approved
- ASTM D6978 Chemotherapy drug tested
Techniglove products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel. Exclusive on-line batch testing is available for this product.
They are approved for use in both USP797/800 and tested per ASTM D6978 for us in chemotherapy applications. Each case is marked with batch numbers to meet internal QC requirements and to ensure full product traceability.
Certifications: COC available on request. Certificate of Sterility available on request.
Ordering information: 200 pair per case. Packaged 1 pair per wallet, 50 wallets per master bag, 4 master bags per case.
Spécifications
- Catalog No:
- 89415-125
- 89415-127
- 89415-121
- 89415-123
- 89415-119
- 89415-117
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous