TGL1400 Series Class 100 Cleanroom Latex Glove, TechNiGlove
Fournisseur: Techniglove
Certificats
A propos de cet article
TechniCleaned ambidextrous 12" Class 100 powder-free latex gloves are designed for controlled environments requiring stringent low particles and extractables.
- Class 100
- 12" ambidextrous
- Powder-free
Techniglove products are made from th highest grade latex available. They contain no silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel. Exclusive On-line Batch Testing is available for this product.
Certifications: COC available on request.
Ordering information: Packaged 100 gloves per poly-sealed cleanroom bag, 10 bags per case.
Spécifications
- Catalog No:
- 89415-090
- 89415-096
- 89415-094
- 89415-092
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Finish:
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips