TN300W Series Class 100 Nitrile Cleanroom Glove, TechNiGlove
Fournisseur: Techniglove
Certificats
A propos de cet article
TechNiGlove Class 100 9.5" ambidextrous powder-free nitrile gloves are puncture-resistant and 100% nitrile, reducing the potential for allergic reactions to natural rubber proteins.
- Class 100
- 9.5" powder-free
- 100% Nitrile
- Ambidextrous
Techniglove products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel. Exclusive On-line Batch Testing is available for this product.
Certifications: COC available on request.
Ordering information: Packaged 100 gloves per poly-sealed cleanroom bag, 10 bags per case.
Spécifications
- Catalog No:
- 89415-082
- 89415-080
- 89415-086
- 89415-084
- 89415-078
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Finish:
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips