TechNiPak Class 100 Nitrile Cleanroom Gloves, TechNiGlove
Fournisseur: Techniglove
Certificats
A propos de cet article
The TechniPak is the world’s only Plastic (PVC) dispenser box of powder-free, 9.5", Class 100 gloves for controlled environments.
- Plastic (PVC) dispenser box
- Class 100
- 9.5" length, powder-free
- 100% Nitrile
- Ambidextrous
Techniglove products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel. Exclusive On-line Batch Testing is available for this product.
The clean PVC box reduces particulate from paper boxes and can be wiped clean in wet applications.
Certifications: COC available on request.
Ordering information: Packaged 100 gloves per TechNiPak PVC box, 10 boxes per case.
Spécifications
- Catalog No:
- 89415-068
- 89415-066
- 89415-076
- 89415-074
- 89415-072
- 89415-070
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Finish:
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips