TechNiTote Class 100 Nitrile Cleanroom Glove, TechNiGlove
Fournisseur: Techniglove
Certificats
A propos de cet article
The TechniTote is a plastic clamshell dispenser containing powder-free, ambidextrous, 12", Class 100 nitrile gloves for use in applications requiring a clean, portable dispenser.
- Plastic (PVC) dispenser case
- Class 100
- 12"" powder-free
- 100% Nitrile
- Ambidextrous
Techniglove products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel. Exclusive On-line Batch Testing is available for this product.
Certifications: COC available on request.
Ordering information: Packaged 250 gloves per TechNiTote, 4 TechNiTotes per case.
Spécifications
- Catalog No:
- 89415-060
- 89415-064
- 89415-062
- 89415-054
- 89415-058
- 89415-056
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Finish:
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips