Corning® 96-Well Clear Round Bottom Polypropylene Microplate, Untreated, Corning
Catalog # 71000-078
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Well Volume:330 µL
- Description:Polypropylene Round-Bottom Plate with Lid
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Cat. no.:71000-078
- Packaging:1/Tray
- Working Volume:75-200 µl
- Well Area:0.32 cm2
- Supplier No.:3879
Specifications
About this item
PS, clear, with lid.
- Corning® CellBIND® surface increases wetability for more even and consistent cell attachment
- Ultra-Low Attachment plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation
- Sterilized by gamma irradiation
- Certified non-pyrogenic
Recommended working volume: 75 to 200 µl.
Frequently Bought Together