Legato 110 DRS Dual Rate Syringe Pump System, KD Scientific
# de catalogue 10117-548
Fournisseur: KD Scientific
cUL Listed
CSA Certified
UL Listed
Spécifications
- Accuracy:± 0.5%
- Max. flow rate:88.28 ml/min
- Min. flow rate:1.26 pl/min
- Description:Legato 110 DRS Dual Rate Syrigne Pump System
- Electrical:100-220V, 50-60 Hz
- Supplier No.:788110DRS
Spécifications
A propos de cet article
This syringe pump system allows the user to configure two different Legato 110 Infuse / Withdraw Programmable pumps with independent flow rates using the built in multistep programming and input / output signal communication
- High resolution color touch screen, touch pad lock feature, LED light on front panel, built in RS285 interface, USB port
- Full metal chassis, built in syringe table
- Up to 30 lb. linear force
- Advanced microstepping techniques
- I/O and TTL interface, continuous mode of operation
- Spill dam, programmable
All control is set through the innovative Legato touch screen user interface. No external computer is required. Accomodates syringe sizes 0.5ul to 60 ml. A communication cable for the two pumps is included.
KD Scientific syringe pumps are for laboratory use only. Not for clinical use.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.
Customers Who Bought This Also Bought