Legato 210 Dual Syringe Infuse/Withdraw Programmable Syringe Pump, KD Scientific
# de catalogue 10117-500
Fournisseur: KD Scientific
cUL Listed
CSA Certified
UL Listed
Spécifications
- Accuracy:± 0.35%
- Max. flow rate:105 ml/min
- Min. flow rate:3.06 pl/min
- Description:Legato 210 Dual Syringe Infuse/Withdraw Programmable Syringe Pump
- Electrical:100-220V, 50-60 Hz
- Supplier No.:788212
Spécifications
A propos de cet article
Infuse/Withdraw/Continuous dual syringe pump (syringes size 0.5ul to 140ml)
- Touch pad lock feature, LED light on front panel, USB port, I/O and TTL interface
- Full metal chassis
- Built in syringe table, RS485 interface
- Advanced microstepping techniques
- Continuous mode of operation
- Spill dam
- Programmable
High resolutuion touch screen display. User definable flow rates with selectable target volume or time values. Programmable with 40 multi-step programs with constant rate, ramp, pulsed & stepped profiles.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.
Customers Who Bought This Also Bought