Legato 130 Single Syringe Infuse/Withdraw Programmable Syringe Pump, KD Scientific
# de catalogue 10117-494
Fournisseur: KD Scientific
cUL Listed
CSA Certified
UL Listed
Spécifications
- Accuracy:± 0.5%
- Max. flow rate:3.818 ml/min
- Min. flow rate:3.66 pl/min
- Description:Legato 130 Single Syringe Infuse/Withdraw Programmable Syringe Pump
- Electrical:100-220V, 50-60 Hz
- Supplier No.:788130
Spécifications
A propos de cet article
Single syringe infuse/ withdraw with program option for 2 programs
- Touch pad lock feature, LED light on front panel, USB port, I/O and TTL interface
- Full metal chassis
- Built in syringe table, RS485 interface
- Advanced microstepping techniques
- Continuous mode of operation
- Spill dam
- Programmable
50 steps each. Holds one syringes from 0.5µl to 1000µl. User definable flow rates with selectable target volume or time values to control the total infusion volume.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.
Customers Who Bought This Also Bought