VWR® Mechanical Cryogenic Freezer Boxes with Dividers
Fournisseur: VWR International
Certificats
A propos de cet article
These Mechanical Cryogenic Freezer Boxes with dividers are constructed of durable fiberboard with a protective moisture repellent coating.
Two and three inch high boxes are available. VWR® Freezer Boxes can be used with most manufacturer's upright and chest freezer racks in general purpose and ultra low mechanical freezers.
Customers Who Bought This Also Bought