Synthware Gooch Crucibles, Low Form, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Low form Gooch Crucible manufactured from synthware glass.
- Heavy wall borosiliacte construction
- Low form style
Available in two porosities, medium or coarse.
Customers Who Bought This Also Bought