Synthware Micro Funnel, Kemtech America
Catalog # 60004-214
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Micro Funnel
- Cat. no.:60004-214
- Top O.D.:27 mm
- Supplier No.:F740027
Specifications
About this item
Micro funnel
- Microscale
- Heavy wall boroscilicate construction
27mm O.D. on top.
Customers Who Bought This Also Bought