Synthware 'Y' Shaped Tubing Connectors
# de catalogue 60004-918
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Synthware 'Y' shaped tubing connectors
- Fits Tubing O.D.:8 mm
- Supplier No.:T110800
Spécifications
A propos de cet article
The 'Y' shaped tubing connectors are manufactured from Synthware glass.
- 8 mm tube connector
- Heavy wall borosilicate glass
Y-shaped connector is ideal for adding another branch to your setup.
Customers Who Bought This Also Bought