Synthware 'T' Shaped Tubing Connectors, Kemtech America
Fournisseur: Kemtech America
Certificats
A propos de cet article
The "T" shaped tubing connectors are manufactured from Synthware glass.
- 8 mm tube connector
- Heay wall borosilicate glass
T-Shaped connector allows for an inline tubing addition.
Customers Who Bought This Also Bought