Stacking Bin Sets
Fournisseur: Treston
Certificats
A propos de cet article
These high quality, durable stacking bins have been designed for daily use in stores, production facilities and warehouses.
- Bins of the same width stack securely, even if they are of different depths
- Straight sides ensure an efficient use of space
- Bright colours make finding the right item easy
- Open front provides easy access
- Corrugated base helps retrieve contents, especially smaller sized and flat objects
- Multiple colors available
The design, with a half-open front, allows excellent visibility and easy access to the bin's contents, making it easy to pick from. The colour options allow a storage system to be designed to meet the customer's requirements.
Desgined to facilitate easy stock selection and picking. Stacking bins are suitable for stands, trolleys, bin rails, table stand, standard metric shelving, and parts retrieval systems.
Ordering information: Labels and dividers sold separately. Installation is required. Assembly videos available.
Customers Who Bought This Also Bought