LED Light Fixtures
Fournisseur: Avantor
Certificats
A propos de cet article
Height and depth adjustable light and balance rails are fitted to the upright tubes and used for suspending lighting units and power tools.
- Maximum load: 33 lbs.
- Can hang tools up to 3 lbs.
Ordering information: Installation is required. Assembly videos available.
Customers Who Bought This Also Bought