Plastic Trays
# de catalogue 470147-486
Fournisseur: Dynalon
Spécifications
- Length:131/4"
- Width:111/4"
- Description:Plastic Trays
- Height:17/8"
- Cat. no.:470147-486
- Supplier No.:921
Spécifications
A propos de cet article
Tray features a pour lip.
HDPE construction
Easily cleaned
Ideal for pond water studies
Customers Who Bought This Also Bought